Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOX14
Entrez Gene ID
504604
Gene Name
SRY-box transcription factor 14
Gene Aliases
-
Chromosome Location
Chr.1: 131371915 - 131372637 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163781.1 NP_001157253.1 1 - 1 622 76

View Full Product Details