Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK26
Entrez Gene ID
539445
Gene Name
serine/threonine kinase 26
Gene Aliases
MST4
Chromosome Location
Chr.X: 15912813 - 15979088 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163786.1 NP_001157258.1 8 - 9 1148 90
GenBank mRNA BC123560.1 - 8 - 9 1148 90

View Full Product Details