Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PURA
Entrez Gene ID
782746
Gene Name
purine rich element binding protein A
Gene Aliases
-
Chromosome Location
Chr.7: 51292891 - 51308498 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205434.1 NP_001192363.1 1 - 2 198 80

View Full Product Details