Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM7
Entrez Gene ID
539804
Gene Name
tripartite motif containing 7
Gene Aliases
-
Chromosome Location
Chr.7: 40326864 - 40341418 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205446.2 NP_001192375.1 5 - 6 1018 103

View Full Product Details