Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT24
Entrez Gene ID
788424
Gene Name
keratin 24
Gene Aliases
-
Chromosome Location
Chr.19: 40902849 - 40907225 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205461.1 NP_001192390.1 8 - 9 1474 69

View Full Product Details