Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMC1
Entrez Gene ID
100140797
Gene Name
regulator of MON1-CCZ1
Gene Aliases
C24H18orf8, WDR98
Chromosome Location
Chr.24: 33105334 - 33127532 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205470.1 NP_001192399.1 6 - 7 687 67
GenBank mRNA BC151667.1 - 6 - 7 659 67

View Full Product Details