Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUAK1
Entrez Gene ID
519892
Gene Name
NUAK family kinase 1
Gene Aliases
-
Chromosome Location
Chr.5: 69476203 - 69552137 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205496.1 NP_001192425.1 2 - 3 599 83

View Full Product Details