Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IHO1
Entrez Gene ID
520504
Gene Name
interactor of HORMAD1 1
Gene Aliases
CCDC36
Chromosome Location
Chr.22: 50812881 - 50840921 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205523.2 NP_001192452.2 4 - 5 435 120
GenBank mRNA BC111307.1 - 4 - 5 436 120

View Full Product Details