Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPAG5
Entrez Gene ID
504585
Gene Name
sperm associated antigen 5
Gene Aliases
-
Chromosome Location
Chr.19: 20034503 - 20053527 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205549.1 NP_001192478.1 23 - 24 3541 91
GenBank mRNA BC149757.1 - 23 - 24 3446 91

View Full Product Details