Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAAL1
Entrez Gene ID
505038
Gene Name
serum amyloid A like 1
Gene Aliases
-
Chromosome Location
Chr.15: 34402569 - 34426197 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205561.1 NP_001192490.1 11 - 12 1330 94

View Full Product Details