Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA24
Entrez Gene ID
506099
Gene Name
spermatogenesis associated 24
Gene Aliases
-
Chromosome Location
Chr.7: 50670489 - 50676007 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205583.1 NP_001192512.1 3 - 4 337 71

View Full Product Details