Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK4
Entrez Gene ID
507800
Gene Name
kallikrein related peptidase 4
Gene Aliases
KLNB1
Chromosome Location
Chr.18: 56902268 - 56906279 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205621.1 NP_001192550.1 2 - 3 221 60

View Full Product Details