Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
508577
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr.28: 29413416 - 29425910 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205641.1 NP_001192570.1 5 - 6 1593 82
GenBank mRNA BC149820.1 - 6 - 7 1669 82

View Full Product Details