Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO30
Entrez Gene ID
539435
Gene Name
F-box protein 30
Gene Aliases
-
Chromosome Location
Chr.9: 83063376 - 83085477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205667.1 NP_001192596.1 1 - 2 2035 83

View Full Product Details