Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP4
Entrez Gene ID
510315
Gene Name
UTP4 small subunit processome component
Gene Aliases
CIRH1A
Chromosome Location
Chr.18: 36343458 - 36373613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205672.1 NP_001192601.1 14 - 15 1703 63

View Full Product Details