Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP55
Entrez Gene ID
513594
Gene Name
centrosomal protein 55
Gene Aliases
-
Chromosome Location
Chr.26: 14823981 - 14845399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205761.3 NP_001192690.1 7 - 8 1338 122

View Full Product Details