Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM15
Entrez Gene ID
540074
Gene Name
tripartite motif containing 15
Gene Aliases
-
Chromosome Location
Chr.23: 28802708 - 28815062 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205764.1 NP_001192693.1 5 - 6 832 74

View Full Product Details