Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRAMP1
Entrez Gene ID
520515
Gene Name
cramped chromatin regulator homolog 1
Gene Aliases
CRAMP1L
Chromosome Location
Chr.25: 1244372 - 1289189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205874.1 NP_001192803.1 19 - 20 3664 91

View Full Product Details