Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35E4
Entrez Gene ID
523783
Gene Name
solute carrier family 35 member E4
Gene Aliases
-
Chromosome Location
Chr.17: 69601336 - 69607796 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205905.3 NP_001192834.1 1 - 2 616 59

View Full Product Details