Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMS2
Entrez Gene ID
527039
Gene Name
PMS1 homolog 2, mismatch repair system component
Gene Aliases
-
Chromosome Location
Chr.25: 38006833 - 38030341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205938.1 NP_001192867.1 13 - 14 2281 87

View Full Product Details