Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GZMK
Entrez Gene ID
528871
Gene Name
granzyme K
Gene Aliases
-
Chromosome Location
Chr.20: 24073111 - 24083866 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205959.1 NP_001192888.1 4 - 5 695 63

View Full Product Details