Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC19A2
Entrez Gene ID
532860
Gene Name
solute carrier family 19 member 2
Gene Aliases
-
Chromosome Location
Chr.16: 37095531 - 37128086 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206010.3 NP_001192939.1 4 - 5 1217 85

View Full Product Details