Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC22
Entrez Gene ID
613943
Gene Name
zinc finger DHHC-type palmitoyltransferase 22
Gene Aliases
-
Chromosome Location
Chr.10: 88330023 - 88340267 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206158.2 NP_001193087.1 1 - 2 532 65

View Full Product Details