Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHIC1
Entrez Gene ID
615065
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
-
Chromosome Location
Chr.X: 77312352 - 77359558 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206195.1 NP_001193124.1 3 - 4 660 101

View Full Product Details