Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM50
Entrez Gene ID
616685
Gene Name
tripartite motif containing 50
Gene Aliases
-
Chromosome Location
Chr.25: 33808726 - 33816136 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206256.3 NP_001193185.1 4 - 5 743 103

View Full Product Details