Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5M
Entrez Gene ID
616992
Gene Name
5',3'-nucleotidase, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.19: 34857016 - 34882469 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206262.1 NP_001193191.1 1 - 2 389 77

View Full Product Details