Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEMD2
Entrez Gene ID
518023
Gene Name
LEM domain nuclear envelope protein 2
Gene Aliases
-
Chromosome Location
Chr.23: 7898378 - 7914749 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192479.1 NP_001179408.1 1 - 2 739 84

View Full Product Details