Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HS6ST1
Entrez Gene ID
518563
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases
-
Chromosome Location
Chr.2: 4187388 - 4224646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192491.1 NP_001179420.1 1 - 2 530 100

View Full Product Details