Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PM20D2
Entrez Gene ID
521179
Gene Name
peptidase M20 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.9: 60958094 - 60971066 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192533.1 NP_001179462.1 2 - 3 602 85

View Full Product Details