Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRE11
Entrez Gene ID
525093
Gene Name
MRE11 homolog, double strand break repair nuclease
Gene Aliases
MRE11A
Chromosome Location
Chr.15: 1434819 - 1508823 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192605.1 NP_001179534.1 16 - 17 2075 71

View Full Product Details