Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD3
Entrez Gene ID
525908
Gene Name
low density lipoprotein receptor class A domain containing 3
Gene Aliases
-
Chromosome Location
Chr.15: 66431232 - 66703031 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192619.1 NP_001179548.1 1 - 2 52 69

View Full Product Details