Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC9
Entrez Gene ID
526819
Gene Name
kelch domain containing 9
Gene Aliases
-
Chromosome Location
Chr.3: 8389186 - 8391006 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192635.1 NP_001179564.1 3 - 4 1012 96

View Full Product Details