Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOP2
Entrez Gene ID
527835
Gene Name
otopetrin 2
Gene Aliases
-
Chromosome Location
Chr.19: 56490907 - 56499714 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192647.3 NP_001179576.3 2 - 3 451 73

View Full Product Details