Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7H5orf46
Entrez Gene ID
614113
Gene Name
chromosome 7 C5orf46 homolog
Gene Aliases
-
Chromosome Location
Chr.7: 59134812 - 59146884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206334.1 NP_001193263.1 3 - 4 313 89
GenBank mRNA BC102132.1 - 3 - 4 315 89
EV681338.1 - 3 - 4 317 89

View Full Product Details