Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERF2
Entrez Gene ID
527852
Gene Name
small EDRK-rich factor 2
Gene Aliases
-
Chromosome Location
Chr.21: 55412552 - 55416413 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206348.1 NP_001193277.1 1 - 2 94 100
GenBank mRNA EV796457.1 - 1 - 2 97 100

View Full Product Details