Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK6
Entrez Gene ID
529814
Gene Name
protein tyrosine kinase 6
Gene Aliases
-
Chromosome Location
Chr.13: 54128175 - 54134496 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192682.1 NP_001179611.1 5 - 6 836 107

View Full Product Details