Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRX5
Entrez Gene ID
532896
Gene Name
iroquois homeobox 5
Gene Aliases
-
Chromosome Location
Chr.18: 23235250 - 23238468 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192744.1 NP_001179673.1 2 - 3 655 72

View Full Product Details