Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIC3
Entrez Gene ID
541035
Gene Name
Zic family member 3
Gene Aliases
-
Chromosome Location
Chr.X: 21146190 - 21149544 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192813.1 NP_001179742.1 2 - 3 1211 85

View Full Product Details