Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LATS1
Entrez Gene ID
535935
Gene Name
large tumor suppressor kinase 1
Gene Aliases
-
Chromosome Location
Chr.9: 86860562 - 86897588 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192866.1 NP_001179795.1 5 - 6 3077 97

View Full Product Details