Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOST
Entrez Gene ID
282880
Gene Name
sclerostin
Gene Aliases
-
Chromosome Location
Chr.19: 43645933 - 43649198 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166514.2 NP_001159986.1 1 - 2 264 82
GenBank mRNA AF326738.1 - 1 - 2 135 82

View Full Product Details