Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKDCC
Entrez Gene ID
539467
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
-
Chromosome Location
Chr.11: 24464835 - 24474942 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206721.1 NP_001193650.1 6 - 7 1561 73
GenBank mRNA EH181331.1 - 5 - 6 661 73

View Full Product Details