Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN7
Entrez Gene ID
539940
Gene Name
UBX domain protein 7
Gene Aliases
UBXD7
Chromosome Location
Chr.1: 70927372 - 70987463 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206764.2 NP_001193693.1 7 - 8 734 65
GenBank mRNA EE353771.1 - 6 - 7 659 65

View Full Product Details