Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHGR1
Entrez Gene ID
787749
Gene Name
proline/histidine/glycine-rich 1
Gene Aliases
-
Chromosome Location
Chr.10: 35966340 - 35973542 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206783.1 NP_001193712.1 1 - 2 37 58
GenBank mRNA EV681842.1 - 1 - 2 37 58

View Full Product Details