Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
VASH1
Entrez Gene ID
615419
Gene Name
vasohibin 1
Gene Aliases
-
Chromosome Location
Chr.10: 88016593 - 88039635 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206803.1 NP_001193732.1 4 - 5 650 53
GenBank mRNA DT899736.1 - 4 - 5 300 53

View Full Product Details