Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNV1
Entrez Gene ID
505732
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
-
Chromosome Location
Chr.14: 54486976 - 54498196 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166560.1 NP_001160032.1 2 - 3 1190 60
GenBank mRNA BC120369.1 - 2 - 3 997 60

View Full Product Details