Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAMP5
Entrez Gene ID
508510
Gene Name
secretory carrier membrane protein 5
Gene Aliases
-
Chromosome Location
Chr.21: 33697299 - 33718476 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166565.1 NP_001160037.1 4 - 5 470 70
GenBank mRNA BC118386.1 - 4 - 5 472 70

View Full Product Details