Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPPB
Entrez Gene ID
508734
Gene Name
natriuretic peptide B
Gene Aliases
-
Chromosome Location
Chr.16: 41822872 - 41824259 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166570.1 NP_001160042.1 2 - 3 387 109

View Full Product Details