Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SPDYA
Entrez Gene ID
616408
Gene Name
speedy/RINGO cell cycle regulator family member A
Gene Aliases
-
Chromosome Location
Chr.11: 70928398 - 70967857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191147.2 NP_001178076.2 4 - 5 486 138
GenBank mRNA BC109600.1 - 4 - 5 470 138
DT833851.1 - 4 - 5 488 138

View Full Product Details