Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IPP
Entrez Gene ID
785293
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
-
Chromosome Location
Chr.3: 100332792 - 100369992 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191196.3 NP_001178125.1 2 - 3 367 126

View Full Product Details