Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBAK
Entrez Gene ID
789791
Gene Name
RB associated KRAB zinc finger
Gene Aliases
-
Chromosome Location
Chr.25: 38535911 - 38549432 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191256.1 NP_001178185.1 2 - 3 553 60

View Full Product Details